Learn More
Medchemexpress LLC BHQ-2 NHS | 916753-62-3 | 97.7% | 603.58 | 50 MG
SDP
Catalog No. 5000404409
Encompass_Preferred
BHQ-2 NHS is a dark quencher with no native emission due to the polyaromatic-azo backbone and a terminal NHS ester. It has a wide and intense quenching range from 560-670 nm, making it useful as an acceptor in fluorescence resonance energy transfer (FRET) applications with orange to far-red emitting dyes. The NHS ester can be used to label primary amines (-NH2) of proteins, amine-modified oligonucleotides, and other amine-containing molecules.
  • Dark quencher with no native emission due to the polyaromatic-azo backbone and a terminal NHS ester.
  • Wide and intense quenching range from 560-670 nm.
  • Useful as an acceptor in fluorescence resonance energy transfer (FRET) applications in conjunction with orange to far-red emitting dyes.
  • NHS ester can be applied to label the primary amines (-NH2) of proteins, amine-modified oligonucleotides, and other amine-containing molecules.
Catalog No. 50-004-04409 Supplier Medchemexpress LLC Supplier No. HY4298450MG
BHQ-2 NHS is a dark quencher with no native emission due to the polyaromatic-azo backbone and a terminal NHS ester. It has a wide and intense quenching range from 560-670 nm, making it useful as an acceptor in fluorescence resonance energy transfer (FRET) applications with orange to far-red emitting dyes. The NHS ester can be used to label primary amines (-NH2) of proteins, amine-modified oligonucleotides, and other amine-containing molecules.
  • Dark quencher with no native emission due to the polyaromatic-azo backbone and a terminal NHS ester.
  • Wide and intense quenching range from 560-670 nm.
  • Useful as an acceptor in fluorescence resonance energy transfer (FRET) applications in conjunction with orange to far-red emitting dyes.
  • NHS ester can be applied to label the primary amines (-NH2) of proteins, amine-modified oligonucleotides, and other amine-containing molecules.