Learn More
Selleck Chemical LLC BHQ 25mg 88-58-4 2,5-di-t-butyl-1,4-benzohydroquinone
SDP
Supplier:  Selleck Chemical LLC S362825MG
Encompass_Preferred
BHQ (2,5-di-t-butyl-1,4-benzohydroquinone) is a potent and selective inhibitor of the sarco-endoplasmic reticulum Ca2+-ATPase (SERCA). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8268