Learn More
LGC GENOMICS LLC BHQ-3 Succinimidyl Ester, 5 mg
Catalog No. NC0838108
Encompass
BHQ-3 OSu is a dark quencher active ester for coupling to amine groups on peptide substrates.
Catalog No. NC0838108 Supplier LGC GENOMICS LLC Supplier No. BHQ3000S5
BHQ-3 OSu is a dark quencher active ester for coupling to amine groups on peptide substrates.