Learn More
U.S. Pharmacopeia Butylated hydroxytoluene, 128-37-0, MFCD00011644, 500 mg
Catalog No. NC9769783
Encompass
Linear Formula: [(CH3)3C]2C6H2(CH3)OH, Molecular Weight: 220.35, BP: 265 °C, MP: 69-73 °C, Synonym: 2,6-Di-tert-butyl-4-methylphenol, 2,6-Di-tert-butyl-p-cresol, BHT, Butylated hydroxytoluene, Butylhydroxytoluene, DBPC.
Catalog No. NC9769783 Supplier U.S. Pharmacopeia Supplier No. 1082708
Linear Formula: [(CH3)3C]2C6H2(CH3)OH, Molecular Weight: 220.35, BP: 265 °C, MP: 69-73 °C, Synonym: 2,6-Di-tert-butyl-4-methylphenol, 2,6-Di-tert-butyl-p-cresol, BHT, Butylated hydroxytoluene, Butylhydroxytoluene, DBPC.