Learn More
Medchemexpress LLC Bi-0115 | 4929-23-1 | 99.7% | 287.74 | 25 MG
SDP
Catalog No. 5000402265
Encompass_Preferred
BI-0115 is a selective inhibitor of LOX-1 (IC50=5.4 μM) that blocks cellular uptake of oxLDL. Its binding triggers receptor inhibition by forming dimers of the homodimeric ligand binding domain.
  • Selective inhibitor of LOX-1.
  • Blocks cellular uptake of oxLDL.
  • Binding triggers receptor inhibition by formation of dimers of the homodimeric ligand binding domain.
  • Efficiently reduces oxLDL internalization in the cellular context with IC50 values (5.4 μM) in good agreement with determined Kd values in SPR (4.3 μM) and ITC (6.99 μM).
Catalog No. 50-004-02265 Supplier Medchemexpress LLC Supplier No. 25MGHY23999
BI-0115 is a selective inhibitor of LOX-1 (IC50=5.4 μM) that blocks cellular uptake of oxLDL. Its binding triggers receptor inhibition by forming dimers of the homodimeric ligand binding domain.
  • Selective inhibitor of LOX-1.
  • Blocks cellular uptake of oxLDL.
  • Binding triggers receptor inhibition by formation of dimers of the homodimeric ligand binding domain.
  • Efficiently reduces oxLDL internalization in the cellular context with IC50 values (5.4 μM) in good agreement with determined Kd values in SPR (4.3 μM) and ITC (6.99 μM).