Learn More
Medchemexpress LLC BI-0115 | 4929-23-1 | 99.7% | 287.74 | 50 MG
SDP
Catalog No. 5000401634
Encompass_Preferred
BI-0115 is a selective inhibitor of LOX-1 (IC50=5.4 μM) that blocks cellular uptake of oxLDL. BI-0115 binding triggers receptor inhibition by formation of dimers of the homodimeric ligand binding domain.
  • Selective inhibitor of LOX-1 (IC50=5.4 μM)
  • Blocks cellular uptake of oxLDL
Catalog No. 50-004-01634 Supplier Medchemexpress LLC Supplier No. HY2399950MG
BI-0115 is a selective inhibitor of LOX-1 (IC50=5.4 μM) that blocks cellular uptake of oxLDL. BI-0115 binding triggers receptor inhibition by formation of dimers of the homodimeric ligand binding domain.
  • Selective inhibitor of LOX-1 (IC50=5.4 μM)
  • Blocks cellular uptake of oxLDL