Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bi 01383298 | 2227549-00-8 | 99.3% | 445.34 | C19H19Cl2FN2O3S | 10MG
SDP

Catalog No. 5000239403
Encompass_Preferred

BI01383298 is a small-molecule chemical probe that potently and selectively inhibits the sodium-coupled citrate transporter SLC13A5 (hNaCT). It is used in research to probe citrate transport and SLC13A5 function in human cells and preclinical studies.

  • Potent inhibitor of SLC13A5: IC50 = 56 nM (hSLC13A5 in HEK cells) and 24 nM (HepG2).
  • Highly selective: negligible activity on other SLC13 family members and murine SLC13A5 (IC50 >100 μM).
  • High purity suitable for research use: 99.3% (HPLC).
  • Molecular formula C19H19Cl2FN2O3S; molecular weight 445.34 g/mol.
  • Provided as a small-quantity research solid for in vitro and preclinical applications.

Catalog No. 50-002-39403 Supplier Medchemexpress LLC Supplier No. HY12473810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BI01383298 is a small-molecule chemical probe that potently and selectively inhibits the sodium-coupled citrate transporter SLC13A5 (hNaCT). It is used in research to probe citrate transport and SLC13A5 function in human cells and preclinical studies.

  • Potent inhibitor of SLC13A5: IC50 = 56 nM (hSLC13A5 in HEK cells) and 24 nM (HepG2).
  • Highly selective: negligible activity on other SLC13 family members and murine SLC13A5 (IC50 >100 μM).
  • High purity suitable for research use: 99.3% (HPLC).
  • Molecular formula C19H19Cl2FN2O3S; molecular weight 445.34 g/mol.
  • Provided as a small-quantity research solid for in vitro and preclinical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.