Learn More
Medchemexpress LLC BI-0474 | 2750570-55-7 | 99.8% | 587.74 | 1 ML
Supplier: Medchemexpress LLC HY14827810MM/1ML
BI-0474 is a potent inhibitor of KRASG12C, exhibiting an IC50 value of 7.0 nM for the GDP-KRAS::SOS1 protein-protein interaction. This compound demonstrates significant anti-proliferative activity against NCI-H358 cells carrying the G12C mutation and shows promising anti-tumor efficacy in non-small cell lung cancer xenograft models.
- Potent KRASG12C inhibitor
- IC50 value of 7.0 nM for GDP-KRAS::SOS1 protein-protein interaction
- Anti-proliferative activity against NCI-H358 cells with G12C mutation (EC50 26 nM)
- Anti-tumor activity in non-small cell lung cancer xenograft models