Learn More
Medchemexpress LLC BI-0474 | 2750570-55-7 | 99.8% | 587.74 | 25 MG
SDP
Catalog No. 5000363747
Encompass_Preferred
BI-0474 is a potent KRASG12C inhibitor demonstrating significant activity against NCI-H358 cells with G12C mutations and anti-tumor effects in non-small cell lung cancer xenograft models.
  • Potent KRASG12C inhibitor
  • IC50 of 7.0 nM for GDP-KRAS::SOS1 protein-protein interaction
  • Exhibits anti-proliferative activity against NCI-H358 cells with G12C mutation
  • Demonstrates anti-tumor activity in non-small cell lung cancer xenograft models
Catalog No. 50-003-63747 Supplier Medchemexpress LLC Supplier No. 25MGHY148278
BI-0474 is a potent KRASG12C inhibitor demonstrating significant activity against NCI-H358 cells with G12C mutations and anti-tumor effects in non-small cell lung cancer xenograft models.
  • Potent KRASG12C inhibitor
  • IC50 of 7.0 nM for GDP-KRAS::SOS1 protein-protein interaction
  • Exhibits anti-proliferative activity against NCI-H358 cells with G12C mutation
  • Demonstrates anti-tumor activity in non-small cell lung cancer xenograft models