Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BI-0474 | 2750570-55-7 | 99.8% | 587.74 | 50 MG
SDP

Catalog No. 5000362353
Encompass_Preferred

BI-0474 is a potent KRASG12C inhibitor that demonstrates an IC50 value of 7.0 nM for the GDP-KRAS::SOS1 protein-protein interaction. It exhibits good anti-proliferative activity against NCI-H358 cells carrying the G12C mutation and shows good anti-tumour activity in non-small cell lung cancer xenograft models. Designed by Boehringer Ingelheim, this compound is available for research use only.

  • Potent KRASG12C inhibitor
  • IC50 value of 7.0 nM for GDP-KRAS::SOS1 protein-protein interaction
  • Demonstrates anti-proliferative activity against NCI-H358 cells with G12C mutation
  • Shows anti-tumour activity in non-small cell lung cancer xenograft models
  • For research use only
  • Purity of 99.83%
  • Molecular weight of 587.74
  • Chemical formula is C30H37N9O2S
  • Appears as a white to off-white solid
  • Target: KRAS-SOS1
  • EC50 = 26 nM on NCI-H358 cells (G12C mutant)
  • Soluble in DMSO at 100 mg/mL

Catalog No. 50-003-62353 Supplier Medchemexpress LLC Supplier No. HY14827850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BI-0474 is a potent KRASG12C inhibitor that demonstrates an IC50 value of 7.0 nM for the GDP-KRAS::SOS1 protein-protein interaction. It exhibits good anti-proliferative activity against NCI-H358 cells carrying the G12C mutation and shows good anti-tumour activity in non-small cell lung cancer xenograft models. Designed by Boehringer Ingelheim, this compound is available for research use only.

  • Potent KRASG12C inhibitor
  • IC50 value of 7.0 nM for GDP-KRAS::SOS1 protein-protein interaction
  • Demonstrates anti-proliferative activity against NCI-H358 cells with G12C mutation
  • Shows anti-tumour activity in non-small cell lung cancer xenograft models
  • For research use only
  • Purity of 99.83%
  • Molecular weight of 587.74
  • Chemical formula is C30H37N9O2S
  • Appears as a white to off-white solid
  • Target: KRAS-SOS1
  • EC50 = 26 nM on NCI-H358 cells (G12C mutant)
  • Soluble in DMSO at 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.