Learn More
Selleck Chemical LLC BI-0474-E1356-5MG
SDP
Catalog No. 503137121
Encompass_Preferred
BI-0474 is a potent inhibitor of KRASG12C with an IC50 value of 7 0 nM It inhibits GDP-KRAS SOS1 protein-protein interactions It exhibits anti-proliferative and anti-tumour activity and can be used in cancer research
Catalog No. 50-313-7121 Supplier Selleck Chemical LLC Supplier No. E13565MG
BI-0474 is a potent inhibitor of KRASG12C with an IC50 value of 7 0 nM It inhibits GDP-KRAS SOS1 protein-protein interactions It exhibits anti-proliferative and anti-tumour activity and can be used in cancer research