Learn More
TARGETMOL CHEMICALS INC BI 1015550 5MG
SDP
Catalog No. 502878129
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Nerandomilast (BI 1015550) is an orally active PDE4B inhibitor with IC50 of 7.2 nM. BI 1015550 has potential applications in inflammation, allergic diseases, pulmonary fibrosis and chronic obstructive pulmonary disease (COPD) with a good safety profile. Purity 99.71%
Catalog No. 50-287-8129 Supplier TARGETMOL CHEMICALS INC Supplier No. T720315MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Nerandomilast (BI 1015550) is an orally active PDE4B inhibitor with IC50 of 7.2 nM. BI 1015550 has potential applications in inflammation, allergic diseases, pulmonary fibrosis and chronic obstructive pulmonary disease (COPD) with a good safety profile. Purity 99.71%