Learn More
Selleck Chemical LLC BI 1015550-E1382-25MG
SDP
Catalog No. 503138273
Encompass_Preferred
BI 1015550 is an orally active inhibitor of PDE4B with IC50 of 7 2 nM It has good safety and potential applications in inflammation allergic diseases pulmonary fibrosis and chronic obstructive pulmonary disease (COPD)
Catalog No. 50-313-8273 Supplier Selleck Chemical LLC Supplier No. E138225MG
BI 1015550 is an orally active inhibitor of PDE4B with IC50 of 7 2 nM It has good safety and potential applications in inflammation allergic diseases pulmonary fibrosis and chronic obstructive pulmonary disease (COPD)