Learn More
TARGETMOL CHEMICALS INC BI-1347 10MG
SDP
Catalog No. 502576687
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BI-1347 is a potent selective inhibitor of CDK8/cyclinC (IC50 1 nM). It shows tumor growth inhibition in an in vivo xenograft model. purity: 99%
Catalog No. 50-257-6687 Supplier TARGETMOL CHEMICALS INC Supplier No. T540510MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BI-1347 is a potent selective inhibitor of CDK8/cyclinC (IC50 1 nM). It shows tumor growth inhibition in an in vivo xenograft model. purity: 99%