Learn More
Medchemexpress LLC Bi-1347 | 2163056-91-3 | ≥98.0% | 1 ML
SDP
Catalog No. 5000234201
Encompass_Preferred
Bi-1347 is a potent, selective small-molecule inhibitor of cyclin-dependent kinase 8 (CDK8), reported IC50 ≈ 1-1.1 nM. It has demonstrated antitumoral activity in preclinical models and is used as a research tool to probe CDK8/19 biology. Supplied as a 10 mM solution in DMSO for in vitro studies; research use only.
  • Potent CDK8 inhibitor (IC50 ≈ 1-1.1 nM)
  • Shows antitumoral activity in preclinical models
  • Also active against CDK19 with high selectivity
  • Provided as 10 mM solution in DMSO for in vitro experiments
  • Intended for research use only; not for human therapeutic use
Catalog No. 50-002-34201 Supplier Medchemexpress LLC Supplier No. HY12035010MM 1ML
Bi-1347 is a potent, selective small-molecule inhibitor of cyclin-dependent kinase 8 (CDK8), reported IC50 ≈ 1-1.1 nM. It has demonstrated antitumoral activity in preclinical models and is used as a research tool to probe CDK8/19 biology. Supplied as a 10 mM solution in DMSO for in vitro studies; research use only.
  • Potent CDK8 inhibitor (IC50 ≈ 1-1.1 nM)
  • Shows antitumoral activity in preclinical models
  • Also active against CDK19 with high selectivity
  • Provided as 10 mM solution in DMSO for in vitro experiments
  • Intended for research use only; not for human therapeutic use