Learn More
Selleck Chemical LLC BI-1347 S1058-25mg
SDP
Catalog No. 503108355
Encompass_Preferred
BI-1347 is a small molecule inhibitor of Cyclin-dependent kinase 8(CDK8) with IC50 of 1 1 nM
Catalog No. 50-310-8355 Supplier Selleck Chemical LLC Supplier No. S105825MG
BI-1347 is a small molecule inhibitor of Cyclin-dependent kinase 8(CDK8) with IC50 of 1 1 nM