Learn More
Selleck Chemical LLC BI-1347 S1058-10mM/1mL
SDP
Catalog No. 503111052
Encompass_Preferred
BI-1347 is a small molecule inhibitor of Cyclin-dependent kinase 8(CDK8) with IC50 of 1 1 nM
Catalog No. 50-311-1052 Supplier Selleck Chemical LLC Supplier No. S105810MM/1ML
BI-1347 is a small molecule inhibitor of Cyclin-dependent kinase 8(CDK8) with IC50 of 1 1 nM