Learn More
Selleck Chemical LLC BI-1347 S1058-1g
SDP
Supplier:  Selleck Chemical LLC S10581G
Encompass_Preferred
BI-1347 is a small molecule inhibitor of Cyclin-dependent kinase 8(CDK8) with IC50 of 1 1 nM
Catalog No. 50-311-1053