Learn More
Selleck Chemical LLC BI-1347-5mg
SDP
Supplier:  Selleck Chemical LLC S10585MG
Encompass_Preferred
BI-1347 is a small molecule inhibitor of Cyclin-dependent kinase 8(CDK8) with IC50 of 1.1 nM.
Catalog No. 50-238-9762