Learn More
Medchemexpress LLC HY-120350 5mg Medchemexpress, BI-1347 CAS:2163056-91-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1203505MG
Encompass_Preferred
Medchemexpress, HY-120350 5mg BI-1347 CAS:2163056-91-3 BI-1347 is a potent CDK8 inhibitor extracted from patent WO2017202719A1, product I-003, has an IC 50 of 1.1 nM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2550