Learn More
Medchemexpress LLC HY-121251 5mg Medchemexpress, BI-167107 CAS:1202235-68-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1212515MG
Encompass_Preferred
Medchemexpress, HY-121251 5mg BI-167107 CAS:1202235-68-4 BI-167107 is a high affinity, full agonist that binds to the β2 adrenergic receptor ( β2AR ) with a dissociation constant K d of 84 pM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2556