Learn More
Medchemexpress LLC BI-1808 10mg | 10mg
SDP
Supplier:  Medchemexpress LLC HYP99119210MG
Encompass_Preferred
BI-1808 is a human IgG1 monoclonal antibody that targets TNFR2by blocking interaction of TNFR2 with ligand TNF- confers Fc R-dependent depletion of Treg and mediates expansion of intratumoral CD8 T cells[1]
Catalog No. 50-003-23932