Learn More
Medchemexpress LLC BI-1808 1mg | 1mg
SDP
Catalog No. 5000323852
Encompass_Preferred
BI-1808 is a human IgG1 monoclonal antibody that targets TNFR2by blocking interaction of TNFR2 with ligand TNF- confers Fc R-dependent depletion of Treg and mediates expansion of intratumoral CD8 T cells[1]
Catalog No. 50-003-23852 Supplier Medchemexpress LLC Supplier No. HYP9911921MG
BI-1808 is a human IgG1 monoclonal antibody that targets TNFR2by blocking interaction of TNFR2 with ligand TNF- confers Fc R-dependent depletion of Treg and mediates expansion of intratumoral CD8 T cells[1]