Learn More
Medchemexpress LLC Bi-1935 | 940954-41-6 | 98.0% | 498.46 g/mol | C24H21F3N6O3 | 10MM 1ML

Supplier: Medchemexpress LLC HY12406310MM 1ML
BI-1935 is a chemical probe and potent soluble epoxide hydrolase (sEH) inhibitor used for preclinical research into cardiovascular disease. It is supplied as a 10 mM solution in DMSO or as solid quantities for formulation and assay development, and is provided at high purity for biochemical and cellular studies.
- Potent soluble epoxide hydrolase (sEH) inhibitor.
- Intended for cardiovascular disease research.
- Available as 10 mM solution in DMSO and as solids (5-100 mg; larger sizes by quote).
- Purity 98.0%.
- Chemical formula C24H21F3N6O3; molecular weight 498.46 g/mol.
- Recommended solvent DMSO; solubility up to 100 mg/mL (200.62 mM).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.