Learn More
Medchemexpress LLC BI 224436 | 1155419-89-8 | 99.7% | 442.51 | 50 MG
SDP
Catalog No. 5000387787
Encompass_Preferred
BI 224436 is a novel HIV-1 noncatalytic site integrase inhibitor. It exhibits EC50 values of less than 15 nM against different HIV-1 laboratory strains. Originally designed by Boehringer Ingelheim, this compound is suitable for various laboratory research applications.
  • Novel HIV-1 noncatalytic site integrase inhibitor
  • Effective against different HIV-1 laboratory strains with EC50 values less than 15 nM
  • Displays an additive effect in combination with most approved antiretrovirals
  • Retains full antiviral activity against recombinant viruses encoding INSTI resistance substitutions
  • Exhibits excellent pharmacokinetic profiles in rat, monkey, and dog
Catalog No. 50-003-87787 Supplier Medchemexpress LLC Supplier No. HY1859550MG
BI 224436 is a novel HIV-1 noncatalytic site integrase inhibitor. It exhibits EC50 values of less than 15 nM against different HIV-1 laboratory strains. Originally designed by Boehringer Ingelheim, this compound is suitable for various laboratory research applications.
  • Novel HIV-1 noncatalytic site integrase inhibitor
  • Effective against different HIV-1 laboratory strains with EC50 values less than 15 nM
  • Displays an additive effect in combination with most approved antiretrovirals
  • Retains full antiviral activity against recombinant viruses encoding INSTI resistance substitutions
  • Exhibits excellent pharmacokinetic profiles in rat, monkey, and dog