Learn More
Medchemexpress LLC BI-2493 | 2937344-16-4 | C24H27N7OS | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1537231MG
Encompass_Preferred
BI-2493 is a structural analogue of BI-2865, functioning as a highly selective and orally active pan-KRAS inhibitor. It is capable of attenuating tumor growth and is applicable for cancer disease research.
  • Functions as a highly selective and orally active pan-KRAS inhibitor
  • Attenuates tumor growth
  • Used for cancer diseases research
  • Inhibits the proliferation of KRAS mutant cancer cell lines in a concentration-dependent manner in vitro
  • Exhibits tumor-suppressive activity in xenograft NMRI mice models in vivo
  • Reduces tumor growth in mice carrying KRAS G12C, G12D, G12V, and A146V mutation models without significant toxicity
Catalog No. 50-003-76120