Learn More
Medchemexpress LLC BI-2493 | 2937344-16-4 | C24H27N7OS | 100 MG
SDP
Catalog No. 5000376164
Encompass_Preferred
BI-2493 is a structural analogue of BI-2865, functioning as a highly selective and orally active pan-KRAS inhibitor. It is utilized in cancer research due to its ability to attenuate tumor growth.
  • Highly selective and orally active pan-KRAS inhibitor
  • Attenuates tumor growth
  • Inhibits proliferation of KRAS mutant cancer cell lines
  • Exhibits tumor-suppressive activity in xenograft models
Catalog No. 50-003-76164 Supplier Medchemexpress LLC Supplier No. HY153723100MG
BI-2493 is a structural analogue of BI-2865, functioning as a highly selective and orally active pan-KRAS inhibitor. It is utilized in cancer research due to its ability to attenuate tumor growth.
  • Highly selective and orally active pan-KRAS inhibitor
  • Attenuates tumor growth
  • Inhibits proliferation of KRAS mutant cancer cell lines
  • Exhibits tumor-suppressive activity in xenograft models