Learn More
Medchemexpress LLC BI-2493 | 2937344-16-4 | C24H27N7OS | 25 MG
SDP
Catalog No. 5000374645
Encompass_Preferred
BI-2493 is a structural analogue of BI-2865, acting as a highly selective and orally active pan-KRAS inhibitor. It can attenuate tumor growth and is suitable for cancer disease research.
  • Highly selective pan-KRAS inhibitor
  • Orally active
  • Attenuates tumor growth
  • Suitable for cancer disease research
Catalog No. 50-003-74645 Supplier Medchemexpress LLC Supplier No. 25MGHY153723
BI-2493 is a structural analogue of BI-2865, acting as a highly selective and orally active pan-KRAS inhibitor. It can attenuate tumor growth and is suitable for cancer disease research.
  • Highly selective pan-KRAS inhibitor
  • Orally active
  • Attenuates tumor growth
  • Suitable for cancer disease research