Learn More
Medchemexpress LLC BI-2493 | 2937344-16-4 | C24H27N7OS | 50 MG
SDP
Catalog No. 5000376296
Encompass_Preferred
BI-2493 is a structural analogue of BI-2865 and a highly selective and orally active pan-KRAS inhibitor. It is designed to attenuate tumor growth and is primarily used for cancer diseases research.
  • Highly selective and orally active pan-KRAS inhibitor
  • Attenuates tumor growth
  • Used for cancer diseases research
  • Inhibits proliferation of KRAS mutant cancer cell lines in a concentration-dependent manner
  • Exhibits tumor-suppressive activity in xenograft mouse models
Catalog No. 50-003-76296 Supplier Medchemexpress LLC Supplier No. HY15372350MG
BI-2493 is a structural analogue of BI-2865 and a highly selective and orally active pan-KRAS inhibitor. It is designed to attenuate tumor growth and is primarily used for cancer diseases research.
  • Highly selective and orally active pan-KRAS inhibitor
  • Attenuates tumor growth
  • Used for cancer diseases research
  • Inhibits proliferation of KRAS mutant cancer cell lines in a concentration-dependent manner
  • Exhibits tumor-suppressive activity in xenograft mouse models