Learn More
TARGETMOL CHEMICALS INC BI-2493 5MG
SDP
Catalog No. 502579253
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. BI-2493 is a highly selective pan-KRAS inhibitor and structural analog of BI-2865.BI-2493 exhibits antitumor activity and inhibits tumor cell growth and can be used in the study of cancerous diseases.BI-2493 is a highly selective pan-KRAS inhibitor and structural analog of BI-2865. purity: 99%
Catalog No. 50-257-9253 Supplier TARGETMOL CHEMICALS INC Supplier No. T720615MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. BI-2493 is a highly selective pan-KRAS inhibitor and structural analog of BI-2865.BI-2493 exhibits antitumor activity and inhibits tumor cell growth and can be used in the study of cancerous diseases.BI-2493 is a highly selective pan-KRAS inhibitor and structural analog of BI-2865. purity: 99%