Learn More
Selleck Chemical LLC BI-2493-E1564-25MG
SDP
Catalog No. 503136553
Encompass_Preferred
BI-2493 a structural analogue of BI-2865 is a highly selective inhibitor of pan-KRAS BI-2493 attenuates tumor growth and can be used for cancer research
Catalog No. 50-313-6553 Supplier Selleck Chemical LLC Supplier No. E156425MG
Add to Cart
Add to Cart
Request a Quote
BI-2493 a structural analogue of BI-2865 is a highly selective inhibitor of pan-KRAS BI-2493 attenuates tumor growth and can be used for cancer research