Learn More
TARGETMOL CHEMICALS INC BI 2536 10MG
SDP
Catalog No. 502879244
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. BI2536 is an effective Plk1 inhibitor (IC50 0.83 nM). It has 4- and 11-fold greater selectivity than Plk2 and Plk3. Purity 98.31%
Catalog No. 50-287-9244 Supplier TARGETMOL CHEMICALS INC Supplier No. T617310MG
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. BI2536 is an effective Plk1 inhibitor (IC50 0.83 nM). It has 4- and 11-fold greater selectivity than Plk2 and Plk3. Purity 98.31%