Learn More
Selleck Chemical LLC BI 2536 S1109-1g
SDP
Catalog No. 503108444
Encompass_Preferred
BI-2536 is a potent Plk1 inhibitor with IC50 of 0 83 nM in a cell-free assay BI-2536 inhibits Bromodomain 4 (BRD4) with Kd of 37 nM and potently suppresses c-Myc expression BI-2536 induces apoptosis and attenuates autophagy Phase 2
Catalog No. 50-310-8444 Supplier Selleck Chemical LLC Supplier No. S11091G
BI-2536 is a potent Plk1 inhibitor with IC50 of 0 83 nM in a cell-free assay BI-2536 inhibits Bromodomain 4 (BRD4) with Kd of 37 nM and potently suppresses c-Myc expression BI-2536 induces apoptosis and attenuates autophagy Phase 2