Learn More
Selleck Chemical LLC BI 2536 10mM/1mL 755038-02-9
SDP
Catalog No. 501360925
Encompass_Preferred
BI-2536 is a potent Plk1 inhibitor with IC50 of 0.83 nM in a cell-free assay. BI-2536 inhibits Bromodomain 4 (BRD4) with Kd of 37 nM and potently suppresses c-Myc expression. BI-2536 induces apoptosis and attenuates autophagy. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-0925 Supplier Selleck Chemical LLC Supplier No. S110910MM/1ML
BI-2536 is a potent Plk1 inhibitor with IC50 of 0.83 nM in a cell-free assay. BI-2536 inhibits Bromodomain 4 (BRD4) with Kd of 37 nM and potently suppresses c-Myc expression. BI-2536 induces apoptosis and attenuates autophagy. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.