Learn More
Apexbio Technology LLC BI 2536(Synonyms: BI-2536, BI2536, Plk1 inhibitor BI 2536), 50mg, CAS: 755038-02-9.
SDP
Catalog No. 5000061067
Encompass_Preferred
BI 2536 (CAS number 755038-02-9) is an ATP-competitive inhibitor selectively targeting human polo-like kinase 1 (PLK1) exhibiting an IC50 value of approximately 0 83 nM Comparatively BI 2536 shows substantially less affinity towards other kinases In vitro studies demonstrate that BI 2536 inhibits proliferation of various human tumor cell lines (EC50 values ranging from 2 to 25 nM) and induces G2/M cell cycle arrest accompanied by apoptosis as evidenced in HeLa cervical cancer cells In vivo data from xenograft tumor models indicate that BI 2536 administered intravenously on a weekly or biweekly schedule possesses antitumor efficacy
Catalog No. 50-000-61067 Supplier Apexbio Technology LLC Supplier No. A396550
BI 2536 (CAS number 755038-02-9) is an ATP-competitive inhibitor selectively targeting human polo-like kinase 1 (PLK1) exhibiting an IC50 value of approximately 0 83 nM Comparatively BI 2536 shows substantially less affinity towards other kinases In vitro studies demonstrate that BI 2536 inhibits proliferation of various human tumor cell lines (EC50 values ranging from 2 to 25 nM) and induces G2/M cell cycle arrest accompanied by apoptosis as evidenced in HeLa cervical cancer cells In vivo data from xenograft tumor models indicate that BI 2536 administered intravenously on a weekly or biweekly schedule possesses antitumor efficacy