Learn More
Medchemexpress LLC BI 2536 | 755038-02-9 | 99.9% | 200 MG
SDP
Catalog No. 5000357901
Encompass_Preferred
BI 2536 is a dual PLK1 and BRD4 inhibitor with IC50s of 0.83 nM and 25 nM, respectively. It suppresses IFNB (encoding IFN-β) gene transcription. This compound is for research use only and not sold to patients.
  • Dual PLK1 and BRD4 inhibitor
  • Suppresses IFNB gene transcription
  • For research use only
  • PLK1 IC50: 0.83 nM
  • BRD4 IC50: 25 nM
Catalog No. 50-003-57901 Supplier Medchemexpress LLC Supplier No. HY50698200MG
BI 2536 is a dual PLK1 and BRD4 inhibitor with IC50s of 0.83 nM and 25 nM, respectively. It suppresses IFNB (encoding IFN-β) gene transcription. This compound is for research use only and not sold to patients.
  • Dual PLK1 and BRD4 inhibitor
  • Suppresses IFNB gene transcription
  • For research use only
  • PLK1 IC50: 0.83 nM
  • BRD4 IC50: 25 nM