Learn More
Medchemexpress LLC BI 2536 | 755038-02-9 | 99.9% | 50 MG
SDP
Catalog No. 5000359288
Encompass_Preferred
BI 2536 is a dual inhibitor targeting both PLK1 (Polo-like Kinase 1) and BRD4 (Bromodomain-containing protein 4), with IC50 values of 0.83 nM and 25 nM, respectively. It suppresses IFNB (Interferon-beta) gene transcription and exhibits antiproliferative activity against various human cancer cell lines. This product is intended for research use only.
  • Targets PLK1 and BRD4
  • High potency (IC50 of 0.83 nM for PLK1 and 25 nM for BRD4)
  • Suppresses IFNB gene transcription
  • Demonstrated antiproliferative activity against a wide range of human cancer cell lines
  • Available in solid form and as a 10 mM solution in DMSO
  • Purity of 99.9%
Catalog No. 50-003-59288 Supplier Medchemexpress LLC Supplier No. HY5069850MG
BI 2536 is a dual inhibitor targeting both PLK1 (Polo-like Kinase 1) and BRD4 (Bromodomain-containing protein 4), with IC50 values of 0.83 nM and 25 nM, respectively. It suppresses IFNB (Interferon-beta) gene transcription and exhibits antiproliferative activity against various human cancer cell lines. This product is intended for research use only.
  • Targets PLK1 and BRD4
  • High potency (IC50 of 0.83 nM for PLK1 and 25 nM for BRD4)
  • Suppresses IFNB gene transcription
  • Demonstrated antiproliferative activity against a wide range of human cancer cell lines
  • Available in solid form and as a 10 mM solution in DMSO
  • Purity of 99.9%