Learn More
Medchemexpress LLC BI 2536 | 755038-02-9 | 99.9% | C28H39N7O3 | 1 G
SDP
Catalog No. 5000358316
Encompass_Preferred
BI 2536 is a dual PLK1 and BRD4 inhibitor with IC50s of 0.83 nM and 25 nM, respectively. It suppresses IFNB (encoding IFN-β) gene transcription. It is suitable for laboratory centrifugation and storage.
  • Dual PLK1 and BRD4 inhibitor
  • Suppresses IFNB gene transcription
  • IC50s of 0.83 nM (PLK1) and 25 nM (BRD4)
  • Functions as an auxiliary reagent for cell therapy manufacture (GMP product version)
Catalog No. 50-003-58316 Supplier Medchemexpress LLC Supplier No. HY506981G
BI 2536 is a dual PLK1 and BRD4 inhibitor with IC50s of 0.83 nM and 25 nM, respectively. It suppresses IFNB (encoding IFN-β) gene transcription. It is suitable for laboratory centrifugation and storage.
  • Dual PLK1 and BRD4 inhibitor
  • Suppresses IFNB gene transcription
  • IC50s of 0.83 nM (PLK1) and 25 nM (BRD4)
  • Functions as an auxiliary reagent for cell therapy manufacture (GMP product version)