Learn More
Medchemexpress LLC BI 2536 | 755038-02-9 | 99.9% | C28H39N7O3 | 100 MG
SDP
Catalog No. 5000358064
Encompass_Preferred
BI 2536 is a dual PLK1 and BRD4 inhibitor. It suppresses IFNB (encoding IFN-β) gene transcription and is intended for laboratory research use.
  • Dual PLK1 and BRD4 inhibitor
  • Suppresses IFNB (encoding IFN-β) gene transcription
  • For research use only
Catalog No. 50-003-58064 Supplier Medchemexpress LLC Supplier No. HY50698100MG
BI 2536 is a dual PLK1 and BRD4 inhibitor. It suppresses IFNB (encoding IFN-β) gene transcription and is intended for laboratory research use.
  • Dual PLK1 and BRD4 inhibitor
  • Suppresses IFNB (encoding IFN-β) gene transcription
  • For research use only