Learn More
Medchemexpress LLC BI 2536 | 755038-02-9 | 99.92% | C28H39N7O3 | 1 ML
SDP
Catalog No. 5000359130
Encompass_Preferred
BI 2536 is a dual inhibitor targeting PLK1 and BRD4, with IC50 values of 0.83 nM and 25 nM respectively. It is known to suppress IFNB (encoding IFN-β) gene transcription and induces mitotic arrest and apoptosis in various human cancer cell lines. This compound has been shown to inhibit tumor growth in vivo and exhibits good blood-brain barrier permeability, causing mitotic arrest in glioblastoma-derived neural stem cells but not normal neural stem cells.
  • Inhibits PLK1 and BRD4.
  • Suppresses IFNB gene transcription.
  • Induces mitotic arrest and apoptosis in cancer cells.
  • Inhibits tumor growth in vivo.
  • Exhibits blood-brain barrier permeability.
  • Used in Western blot, immunofluorescence, and growth inhibition assays.
Catalog No. 50-003-59130 Supplier Medchemexpress LLC Supplier No. HY5069810MM/1ML
BI 2536 is a dual inhibitor targeting PLK1 and BRD4, with IC50 values of 0.83 nM and 25 nM respectively. It is known to suppress IFNB (encoding IFN-β) gene transcription and induces mitotic arrest and apoptosis in various human cancer cell lines. This compound has been shown to inhibit tumor growth in vivo and exhibits good blood-brain barrier permeability, causing mitotic arrest in glioblastoma-derived neural stem cells but not normal neural stem cells.
  • Inhibits PLK1 and BRD4.
  • Suppresses IFNB gene transcription.
  • Induces mitotic arrest and apoptosis in cancer cells.
  • Inhibits tumor growth in vivo.
  • Exhibits blood-brain barrier permeability.
  • Used in Western blot, immunofluorescence, and growth inhibition assays.