Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bi-2852 | 2375482-51-0 | 99.5% | 516.59 g/mol | C31H28N6O2 | 100MG
SDP

Catalog No. 5000239650
Encompass_Preferred

BI-2852 is a small-molecule inhibitor that targets the KRAS switch I/II pocket for research applications. It binds the SI/II pocket with nanomolar affinity and disrupts GEF, GAP, and effector interactions, leading to inhibition of downstream KRAS signaling and antiproliferative effects in KRAS-mutant cells.

  • Purity 99.46%.
  • Molecular weight 516.59 g/mol.
  • Chemical formula C31H28N6O2.
  • Soluble in DMSO (55 mg/mL) for in vitro studies.
  • Available as solid and as DMSO solution formats for flexibility in experiments.
  • Storage recommendations: 4°C for solid; in solvent -80°C (2 years) or -20°C (1 year) under nitrogen.

Catalog No. 50-002-39650 Supplier Medchemexpress LLC Supplier No. HY126247100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BI-2852 is a small-molecule inhibitor that targets the KRAS switch I/II pocket for research applications. It binds the SI/II pocket with nanomolar affinity and disrupts GEF, GAP, and effector interactions, leading to inhibition of downstream KRAS signaling and antiproliferative effects in KRAS-mutant cells.

  • Purity 99.46%.
  • Molecular weight 516.59 g/mol.
  • Chemical formula C31H28N6O2.
  • Soluble in DMSO (55 mg/mL) for in vitro studies.
  • Available as solid and as DMSO solution formats for flexibility in experiments.
  • Storage recommendations: 4°C for solid; in solvent -80°C (2 years) or -20°C (1 year) under nitrogen.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.