Learn More
TARGETMOL CHEMICALS INC BI-2852 5MG
SDP
Catalog No. 502581712
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. BI-2852 is a KRAS inhibitor for the switch I/II pocket (SI/II-pocket) with nanomolar affinity. BI-2852 is leading to inhibition of downstream signaling and an antiproliferative effect in KRAS mutant cells. purity: 98%
Catalog No. 50-258-1712 Supplier TARGETMOL CHEMICALS INC Supplier No. T105335MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. BI-2852 is a KRAS inhibitor for the switch I/II pocket (SI/II-pocket) with nanomolar affinity. BI-2852 is leading to inhibition of downstream signaling and an antiproliferative effect in KRAS mutant cells. purity: 98%