Learn More
Medchemexpress LLC BI-2865 | 2937327-93-8 | C23H27N7O2S | 1 MG
SDP
Catalog No. 5000376152
Encompass_Preferred
BI-2865 is a non-covalent pan-KRAS inhibitor. It binds to wild-type (WT) and several mutant KRAS forms, including G12C, G12D, G12V, and G13D, with KDs ranging from 4.3 to 32 nM. This compound effectively inhibits the proliferation of G12C, G12D, or G12V mutant KRAS expressing BaF3 cells, showing a mean IC50 of approximately 140 nM.
  • Binds to multiple KRAS mutants (WT, G12C, G12D, G12V, G13D)
  • Inhibits proliferation of KRAS mutant expressing BaF3 cells
  • Derivative with a prolinol substituent and a pyrimidine linker
  • Exhibits direct ionic interaction with E62 and a water-mediated hydrogen bond network
Catalog No. 50-003-76152 Supplier Medchemexpress LLC Supplier No. HY1537241MG
BI-2865 is a non-covalent pan-KRAS inhibitor. It binds to wild-type (WT) and several mutant KRAS forms, including G12C, G12D, G12V, and G13D, with KDs ranging from 4.3 to 32 nM. This compound effectively inhibits the proliferation of G12C, G12D, or G12V mutant KRAS expressing BaF3 cells, showing a mean IC50 of approximately 140 nM.
  • Binds to multiple KRAS mutants (WT, G12C, G12D, G12V, G13D)
  • Inhibits proliferation of KRAS mutant expressing BaF3 cells
  • Derivative with a prolinol substituent and a pyrimidine linker
  • Exhibits direct ionic interaction with E62 and a water-mediated hydrogen bond network