Learn More
Medchemexpress LLC BI-2865 | 2937327-93-8 | C23H27N7O2S | 1 ML
SDP
Catalog No. 5000376872
Encompass_Preferred
BI-2865 is a non-covalent pan-KRAS Inhibitor. It binds to wild-type and various mutant KRAS forms, including G12C, G12D, G12V, and G13D, inhibiting the proliferation of mutant KRAS expressing BaF3 cells. Characterized by a prolinol substituent and a pyrimidine linker, it forms specific interactions within the KRAS binding site. This product is for research use only.
  • Binds to wild-type and various mutant KRAS forms
  • Inhibits proliferation of mutant KRAS expressing BaF3 cells
  • Features a prolinol substituent and a pyrimidine linker
  • Demonstrates direct ionic interaction with E62
  • Forms a water-mediated hydrogen bond network with R68 and Q61
Catalog No. 50-003-76872 Supplier Medchemexpress LLC Supplier No. HY15372410MM/1ML
BI-2865 is a non-covalent pan-KRAS Inhibitor. It binds to wild-type and various mutant KRAS forms, including G12C, G12D, G12V, and G13D, inhibiting the proliferation of mutant KRAS expressing BaF3 cells. Characterized by a prolinol substituent and a pyrimidine linker, it forms specific interactions within the KRAS binding site. This product is for research use only.
  • Binds to wild-type and various mutant KRAS forms
  • Inhibits proliferation of mutant KRAS expressing BaF3 cells
  • Features a prolinol substituent and a pyrimidine linker
  • Demonstrates direct ionic interaction with E62
  • Forms a water-mediated hydrogen bond network with R68 and Q61