Learn More
Medchemexpress LLC BI-2865 | 2937327-93-8 | C23H27N7O2S | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY153724
Encompass_Preferred
BI-2865 is a non-covalent pan-KRAS inhibitor that binds to wild-type and various mutant KRAS proteins. It effectively inhibits the proliferation of KRAS-expressing cells, making it suitable for research into KRAS-driven cancers and related pathways.
  • Non-covalent pan-KRAS inhibitor
  • Binds to WT, G12C, G12D, G12V, and G13D mutant KRAS
  • Inhibits proliferation of G12C, G12D, or G12V mutant KRAS expressing BaF3 cells
  • High purity (99.65%)
  • Molecular weight: 465.57
  • Derivative with a prolinol substituent and a pyrimidine linker
  • Exhibits direct ionic interaction with E62
  • Forms a water-mediated hydrogen bond network with R68 and Q61
Catalog No. 50-003-75608