Learn More
Selleck Chemical LLC BI-2865-E1474-100MG
SDP
Catalog No. 503134547
Encompass_Preferred
BI-2865 is a none-covalent pan-KRAS Inhibitor It binds to WT G12C G12D G12V and G13D mutant KRAS with KDs of 6 9 4 5 32 26 4 3 nM respectively and inhibits the proliferation of G12C G12D or G12V mutant KRAS expressing BaF3 ce
Catalog No. 50-313-4547 Supplier Selleck Chemical LLC Supplier No. E1474100MG
BI-2865 is a none-covalent pan-KRAS Inhibitor It binds to WT G12C G12D G12V and G13D mutant KRAS with KDs of 6 9 4 5 32 26 4 3 nM respectively and inhibits the proliferation of G12C G12D or G12V mutant KRAS expressing BaF3 ce