Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BI-3231 | 2894848-07-6 | 99.8% | 380.37 | 1 ML
SDP

Catalog No. 5000362360
Encompass_Preferred

BI 3231 is a chemical probe and a potent and selective inhibitor of hydroxysteroid 17β-dehydrogenase 13 (HSD17B13). It has IC50s of 1 nM for hHSD17B13 and 13 nM for mHSD17B13. This compound is being researched for its potential in treating nonalcoholic steatohepatitis (NASH) and other liver diseases.

  • Potent and selective HSD17B13 inhibitor
  • IC50s of 1 nM (hHSD17B13) and 13 nM (mHSD17B13)
  • Potential for research into nonalcoholic steatohepatitis (NASH) and other liver diseases
  • High metabolic stability in liver microsomes
  • Moderate metabolic stability in hepatocytes

Catalog No. 50-003-62360 Supplier Medchemexpress LLC Supplier No. HY14881410MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

BI 3231 is a chemical probe and a potent and selective inhibitor of hydroxysteroid 17β-dehydrogenase 13 (HSD17B13). It has IC50s of 1 nM for hHSD17B13 and 13 nM for mHSD17B13. This compound is being researched for its potential in treating nonalcoholic steatohepatitis (NASH) and other liver diseases.

  • Potent and selective HSD17B13 inhibitor
  • IC50s of 1 nM (hHSD17B13) and 13 nM (mHSD17B13)
  • Potential for research into nonalcoholic steatohepatitis (NASH) and other liver diseases
  • High metabolic stability in liver microsomes
  • Moderate metabolic stability in hepatocytes

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.