Learn More
Medchemexpress LLC BI-3231 | 2894848-07-6 | 99.8% | 380.37 | 5 MG
SDP
Catalog No. 5000359627
Encompass_Preferred
BI 3231 is a potent and selective hydroxysteroid 17β-dehydrogenase 13 (HSD17B13) inhibitor. It has potential for research into nonalcoholic steatohepatitis (NASH) and other liver diseases.
  • Potent and selective HSD17B13 inhibitor
  • IC50s of 1 nM for hHSD17B13 and 13 nM for mHSD17B13
  • Potential for research into nonalcoholic steatohepatitis (NASH)
  • Useful for other liver diseases research
  • Demonstrates high metabolic stability in liver microsomes
  • Moderate metabolic stability in hepatocytes
  • Considerable hepatic exposure maintained over 48 hours
Catalog No. 50-003-59627 Supplier Medchemexpress LLC Supplier No. HY1488145MG
BI 3231 is a potent and selective hydroxysteroid 17β-dehydrogenase 13 (HSD17B13) inhibitor. It has potential for research into nonalcoholic steatohepatitis (NASH) and other liver diseases.
  • Potent and selective HSD17B13 inhibitor
  • IC50s of 1 nM for hHSD17B13 and 13 nM for mHSD17B13
  • Potential for research into nonalcoholic steatohepatitis (NASH)
  • Useful for other liver diseases research
  • Demonstrates high metabolic stability in liver microsomes
  • Moderate metabolic stability in hepatocytes
  • Considerable hepatic exposure maintained over 48 hours