Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BI-3231 | 2894848-07-6 | 99.8% | 380.37 | 50 MG
SDP

Catalog No. 5000361501
Encompass_Preferred

BI-3231 is a potent and selective chemical probe that inhibits hydroxysteroid 17β-dehydrogenase 13 (HSD17B13), with IC50 values of 1 nM for hHSD17B13 and 13 nM for mHSD17B13. It is being researched for its potential in treating nonalcoholic steatohepatitis (NASH) and other liver diseases.

  • Potent and selective hydroxysteroid 17β-dehydrogenase 13 (HSD17B13) inhibitor
  • Demonstrates high metabolic stability in liver microsomes
  • Moderate metabolic stability in hepatocytes
  • Significant hepatic exposure maintained over 48 hours
  • Potential for research into nonalcoholic steatohepatitis (NASH) and other liver diseases
  • Powder storage: -20°C for 3 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Solubility in DMSO: 125 mg/mL (328.63 mM)

Catalog No. 50-003-61501 Supplier Medchemexpress LLC Supplier No. HY14881450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BI-3231 is a potent and selective chemical probe that inhibits hydroxysteroid 17β-dehydrogenase 13 (HSD17B13), with IC50 values of 1 nM for hHSD17B13 and 13 nM for mHSD17B13. It is being researched for its potential in treating nonalcoholic steatohepatitis (NASH) and other liver diseases.

  • Potent and selective hydroxysteroid 17β-dehydrogenase 13 (HSD17B13) inhibitor
  • Demonstrates high metabolic stability in liver microsomes
  • Moderate metabolic stability in hepatocytes
  • Significant hepatic exposure maintained over 48 hours
  • Potential for research into nonalcoholic steatohepatitis (NASH) and other liver diseases
  • Powder storage: -20°C for 3 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Solubility in DMSO: 125 mg/mL (328.63 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.