Learn More
Medchemexpress LLC BI-3231 | 2894848-07-6 | 99.8% | 380.37 | 500 MG
SDP
Catalog No. 5000359628
Encompass_Preferred
BI-3231 is a chemical probe that functions as a potent and selective inhibitor of hydroxysteroid 17β-dehydrogenase 13 (HSD17B13). It exhibits IC50 values of 1 nM for human HSD17B13 and 13 nM for mouse HSD17B13. This compound holds potential for research into nonalcoholic steatohepatitis (NASH) and various other liver diseases. The BI-3231 probe was developed by Boehringer Ingelheim and is available for free through their open innovation portal, opnMe.com, alongside its negative control.
  • Potent and selective HSD17B13 inhibitor.
  • IC50 values: 1 nM (hHSD17B13) and 13 nM (mHSD17B13).
  • Potential for research in nonalcoholic steatohepatitis (NASH) and liver diseases.
  • High metabolic stability in liver microsomes.
  • Moderate metabolic stability in hepatocytes.
  • Rapidly cleared from plasma, with considerable hepatic exposure maintained over 48 hours in vivo.
Catalog No. 50-003-59628 Supplier Medchemexpress LLC Supplier No. HY148814500MG
BI-3231 is a chemical probe that functions as a potent and selective inhibitor of hydroxysteroid 17β-dehydrogenase 13 (HSD17B13). It exhibits IC50 values of 1 nM for human HSD17B13 and 13 nM for mouse HSD17B13. This compound holds potential for research into nonalcoholic steatohepatitis (NASH) and various other liver diseases. The BI-3231 probe was developed by Boehringer Ingelheim and is available for free through their open innovation portal, opnMe.com, alongside its negative control.
  • Potent and selective HSD17B13 inhibitor.
  • IC50 values: 1 nM (hHSD17B13) and 13 nM (mHSD17B13).
  • Potential for research in nonalcoholic steatohepatitis (NASH) and liver diseases.
  • High metabolic stability in liver microsomes.
  • Moderate metabolic stability in hepatocytes.
  • Rapidly cleared from plasma, with considerable hepatic exposure maintained over 48 hours in vivo.